Learn More
PSF Progressive Safety™ Anti-static Safety Wellington Boots To EN345
Click to view available options
Size:
3
4
5
6
7
8
9
10
11
12
13
Unit Size:
1 pair
Description
- Antislip
- Anti-static
- Waterproof rubber sole
- Resistant to heat, oils, acids & animal fats
- 200 joule toe protection
- White
Specifications
Specifications
| Size | 7 |