missing translation for 'onlineSavingsMsg'
Learn More

Stat6 Mouse, Unlabeled, Clone: 23/STAT6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611290

 View more versions of this product

Product Code. 15895859

  • ONLINE EXCLUSIVE PRICE
    £229.00
    £219.86 / 1 set
    Save £9.14
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

STATs (signal transducers and activators of transcription) are critical mediators of the biologic activity of cytokines including Interleukins (IL) 2-5, IL-7, IL-15, GM-CSF, erythropoietin and growth hormone. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of STATs by tyrosine phosphorylation. Active STATs then move to the nucleus to promote transcription of cytokine-inducible genes. Seven STAT proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat6 plays an important role in signaling pathways that lead to the differentiation of T helper type 2 (Th2) cells from uncommitted CD4 T cell precursors. Moreover, IL-4, secreted by activated T lymphocytes, basophils, and mast cells, induces specific gene expression via the induction of tyrosine phosphorylation of Stat6 at tyrosine 641 (Y641). The SH3:SH2 domain of Stat6 associates with tyrosine-phosphorylated IL-4 receptor and the proximal Jak kinase phosphorylates Stat6 at Y641 on the C-terminal side of the SH2 domain. Stat6 is then released from the receptor, dimerizes, and is thought to contact the basal transcription machinery by binding to p300/CBP. While Stat6 is widely expressed in human tissues, it exhibits elevated expression in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung, and placenta.

Host Species: Mouse
Clone: 23
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Stat6 C-terminal Recombinant Protein
Formula Weight [Chemical]: 100kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stat6
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Stat6 C-terminal Recombinant Protein
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Western Blot
23/STAT6
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.