Learn More
Stat5 (pY694) Mouse, PE, Clone: 47, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 612567
Description
Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Seven Stat proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat5 has been characterized and shown to be encoded by two separate genes, Stat5a and Stat5b that share over 90% identity at the amino acid level. Stat5a has been shown to be involved in lactogenesis andmammary development, while Stat5b has been shown to be involved in growth hormone signaling and to play a role in liver gene expression. Both Stat5a and Stat5b share similarities, both are involved in IL-2 induced peripheral T cell proliferation. The peptide hormone, prolactin, binds to the prolactin receptor (PRLR) to initiate the lactogenic response. There are at least three forms of PRLR; however, only the long form is able to activate the 92kDa Stat5 protein by inducing phosphorylation at Y694. Once phosphorylated, Stat5 becomes an essential transcription factor which binds to the β-casein gene promoter. The presence of an SH2 domain within Stat5 suggests that it may directly interact with protein tyrosine kinases (PTKs) such as JAK2.The 47 monoclonal antibody recognizes the phosphorylated Y694 of Stat5a. The homologous phosphorylation site in Stat5b is Y699.
Flow Cytometry (Intracellular Staining)
Specifications
| Stat5 (pY694) | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated Sheep Stat5 Peptide | |
| 50 Tests | |
| Cell Biology | |
| Human, Mouse, Rat, Sheep | |
| IgG1 κ |
| Flow Cytometry | |
| 47 | |
| Signal transducer and activator of transcription 5; MGF; MPF | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.