missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Stat5 (pY694); Clone: 47/Stat5(pY694); BD
Brand: BD Biosciences 562984
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone 47/Stat5(pY694)
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Phosphorylated Sheep Stat5 Peptide
Intracellular block/Flow Cytometry
Specifications
| Stat5 (pY694) | |
| 47/Stat5(pY694) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
| Monoclonal | |
| Brilliant Violet 421 | |
| Phosphorylated Sheep Stat5 Peptide | |
| 50 Tests | |
| Primary | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction