Learn More
Stat3 (pY705) Mouse, Unlabeled, Clone: 4/P-STAT3, BD
Mouse Monoclonal Antibody
£275.79 - £385.31
Specifications
| Antigen | Stat3 (pY705) |
|---|---|
| Clone | 4/P-STAT3 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
10615975
|
BD Biosciences
612356 |
50 μg |
£287.00 £275.79 / Each Save £11.21 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15846179
|
BD Biosciences
612357 |
150 μg |
£401.00 £385.31 / Each Save £15.69 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Seven Stat proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat3 is a 92kDa protein that is activated as a DNA- binding protein through cytokines, such as IL-6, and growth factors, such as EGF. Stat3 activation occurs via tyrosine phosphorylation at Y705. Tyrosine phosphorylation in response to cytokine stimulation is generally mediated by JAK1. Upon activation, Stat3 dimerizes, translocates to the nucleus and binds DNA response elements, thereby regulating gene expression. It has been reported that Stat3 binds to DNA as a homodimer, but it is also capable of binding as a heterodimer with Stat1. In addition to tyrosine phosphorylation, Stat3 is also phosphorylated at S727 via the MAPK pathway. Stat3 is widely expressed and can bind to the sis-inducible element (SIE) site from the c-fos promoter. This site is similar to the GAS element that is present in IFN-γ induced genes. Thus, phosphorylation of Y705 in Stat3 occurs in response to growth factors and cytokines, and is essential for normal transcription activity.
Intracellular Staining, Western Blotting
Specifications
| Stat3 (pY705) | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified |
| 4/P-STAT3 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Phosphorylated Human Stat3 Peptide | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.