Learn More
Stat2 (pY690) Mouse anti-Human, Unlabeled, Clone: 7A/STAT2 (PY690), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558095
Description
Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment/activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Seven Stat proteins have been cloned, each of which is differentially expressed and/or activated in a cytokine-specific and cell type-specific manner. Stat1 and Stat2 are components of the ISGF3 (Interferon-Stimulated Gene Factor 3) complex, which is the primary transcription activator induced by the binding of the interferon to a specific cell-surface receptor. Stat2 is a 113kDa protein having approximately 40% homology with Stat1a. Stat2 interacts with Stat1 for the efficient activation of Stat1 in response to IFN-α, which is essential for normal transcriptional activity of the ISGF3 complex. Stat2 phosphorylation at the tyrosine 690 residue (Y690) after IFN-α activation can occur independently of Stat1, but the localization and nuclear stability of phosphorylated Stat2 is dependent on Stat1. The 7a/Stat2 monoclonal antibody recognizes the phosphorylated Y690 of human Stat2.
Host Species: Mouse
Clone: 7a
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human Stat2 Peptide
Western Blotting
Specifications
| Stat2 (pY690) | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Phosphorylated Human Stat2 Peptide | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgG1 κ |
| Western Blot | |
| 7A/STAT2 (PY690) | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.