missing translation for 'onlineSavingsMsg'
Learn More

Stat1 Mouse, Unlabeled, Clone: 42, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610185

 View more versions of this product

Product Code. 15805319

  • ONLINE EXCLUSIVE PRICE
    £229.00
    £219.86 / 1 set
    Save £9.14
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Stat proteins function as both cytoplasmic signal transducers and activators of transcription. The Stat91/84 (the two proteins are the result of alternate splicing-Stat91 has an additional 38 C-terminal amino acids) and Stat113 were the first identified members of this protein family. These three polypeptides contain both SH2 and SH3 domains and have also been described as members of the ISGF3 (interferon-stimulated gene factor 3) complex. With the discovery of additional members of the Stat family (Stats3 & 4), the nomenclature has been revised to indicate the Stat family members in the order of their discovery. Stat 91, 84, and 113 have become Stat1α, Stat1β, and Stat2, respectively. Stat1α is present in a higher concentration than Stat1β in most cell types. In response to IFNα treatment, Stat1α, Stat1β, and Stat2 become tyrosine-phosphorylated and migrate to the nucleus where they join a 48kDa DNA binding protein and subsequently direct the transcription at IFNα responsive elements. In IFN-γ treated cells, Stat1α (but not Stat2) becomes phosphorylated and forms a dimer. It then enters the nucleus and binds to the IFN-γ activated site (GAS) element in order to direct IFN-γ activated transcription.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stat1
Monoclonal
250μg/mL
C-Terminus
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
42
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.