missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STC2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA534841

Product Code. 15398741

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A549, H1299, MCF-7, Mouse kidney. Predicted reactivity: Mouse (92%), Rat (91%), Pig (96%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

STC2
Polyclonal
Unconjugated
STC2
AW125853; mustc2; stanniocalcin 2; stanniocalcin-2; Stanniocalcin-related protein; STC2; STC-2; Stc2l; STC-related protein; STCRP
Rabbit
Antigen affinity chromatography
RUO
20856, 8614
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O76061, O88452
STC2
Recombinant protein encompassing a sequence within the center region of human Stanniocalcin 2. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.