missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ST2/IL-1 R4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
| Host Species | Rabbit |
|---|---|
| Isotype | IgG |
| Immunogen | A portion of amino acids 25-65 of human ST2 protein was used as the immunogen. |
| Antigen | ST2/IL-1 R4 |
| Primary or Secondary | Primary |
Description
ST2/IL-33R Polyclonal specifically detects ST2/IL-33R in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Rabbit | |
| A portion of amino acids 25-65 of human ST2 protein was used as the immunogen. | |
| Primary | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 9173 | |
| 1.0 mg/mL |
| IgG | |
| ST2/IL-1 R4 | |
| RUO | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA_d) and 94% homologous in chimpanzee. | |
| PBS with 0.05% Sodium Azide |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title