Learn More
anti-SSX2 Matched Antibody Pair, Abnova
Brand: Abnova H00006757-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-SSX2 (100μg); Mouse polyclonal anti-SSX2 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
The product of this gene belongs to the family of highly homologous synovial sarcoma X (SSX) breakpoint proteins. These proteins may function as transcriptional repressors. They are also capable of eliciting spontaneously humoral and cellular immune responses in cancer patients, and are potentially useful targets in cancer vaccine-based immunotherapy. SSX1, SSX2 and SSX4 genes have been involved in the t(X;18) translocation characteristically found in all synovial sarcomas. This translocation results in the fusion of the synovial sarcoma translocation gene on chromosome 18 to one of the SSX genes on chromosome X. The encoded hybrid proteins are probably responsible for transforming activity. Two transcript variants encoding distinct isoforms have been identified for this gene.
ELISA
Specifications
| SSX2 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SSX2. | |
| HD21/HOM-MEL-40/MGC119055/MGC15364/MGC3884/SSX | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-SSX2 (100 μg) 2. Detection antibody: mouse polyclonal anti-SSX2 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| SSX2 | |
| SSX2 | |
| Affinity Purified | |
| RUO | |
| 6757 | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.