missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSTR5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534224
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Marmoset (94%).
Somatostatin is a multifunctional protein affecting exocrine and endocrine protein secretion, neurotransmission, and neuronal modulation and regulating cell proliferation. All these functions of Somatostatin are executed through a class of GPCR protein family members called somatostatin receptor. Till date, 6 classes of somatostatin receptors have been discovered out of which SSTR5, 364A.A protein, is a receptor subtype expressed predominantly in the pituitary and hypothalamus region of brain. Studies revealed that SSTR5 contains seven transmembrane regions and undergoes homodimerization and also heterodimerizes with SSTR1. Although the specific functions of SSTR5 are still elusive, the studies indicate that it complements SSTR2 regulation of ACTH secretion. Polymorphisms in SSTR5 gene have been observed in patients with acromegaly.Specifications
SSTR5 | |
Polyclonal | |
Unconjugated | |
Sstr5 | |
Smstr5; Somato; somatostatin receptor; somatostatin receptor 5; Somatostatin receptor subtype 5; somatostatin receptor type 5; SS5R; SS-5-R; SS5-R; sst5; Sstr5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6755 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P35346 | |
Sstr5 | |
Synthetic 18 amino acid peptide from C-terminus of human SSTR5. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |