Learn More
slingshot homolog 3 (Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SSH3 protein.
Brand: Abnova H00054961-D01P.100ug
Description
The ADF (actin-depolymerizing factor)/cofilin family (see MIM 601442) is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1; MIM 601329). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo (Niwa et al., 2002 [PubMed 11832213]).[supplied by OMIM
Sequence: MALVTVSRSPPGSGASTPVGPWDQAVQRRSRLQRRQSFAVLRGAVLGLQDGGDNDDAAEASSEPTEKAPSEEELHGDQTDFGQGSQSPQKQEEQRQHLHLMVQLLRPQDDIRLAAQLEAPRPPRLRYLLVVSTREGEGLSQDETVLLGVDFPDSSSPSCTLGLVLPLWSDTQVYLDGDGGFSVTSGGQSRIFKPISIQTMWATLQVLHQACEAALGSGLVPGGSALTWASHYQERLNSEQSCLNEWTAMADLESLRPPSAEPGGSSEQEQMEQAIRAELWKVLDVSDLESVTSKEIRQALELRLGLPLQQYRDFIDNQMLLLVAQRDRASRIFPHLYLGSEWNAANLEELQRNRVTHILNMAREIDNFYPERFTYHNVRLWDEESAQLLPHWKETHRFIEAARAQGTHVLVHCKMGVSRSAATVLAYAMKQYECSLEQALRHVQELRPIARPNPGFLRQLQIYQGILTASRQSHVWEQKVGGVSPEEHPAPEVSTPFPPLPPEPEGGGEEKVVGMEESQAAPKEEPGPRPRINLRGVMRSISLLEPSLELESTSETSDMPEVFSSHESSHEEPLQPFPQLARTKGGQQVDRGPQPALKSRQSVVTLQGSAVVANRTQAFQEQEQGQGQGQGEPCISSTPRFRKVVRQASVHDSGEEGEASpecifications
| slingshot homolog 3 (Drosophila) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human SSH3 protein. | |
| SSH3 | |
| FLJ10928/FLJ20515/SSH-3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC007709.1 | |
| SSH3 | |
| SSH3 (AAH07709.1, 1 a.a. ∼ 659 a.a) full-length human protein. | |
| 100 μg | |
| Cell Adhesion | |
| 54961 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.