missing translation for 'onlineSavingsMsg'
Learn More

SSEA-4 Mouse, PerCP-Cy5.5, Clone: MC813-70, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561565

Product Code. 15811139

  • ONLINE EXCLUSIVE PRICE
    £205.00
    £195.85 / Each
    Save £9.15
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The MC813-70 monoclonal antibody reacts with Stage-Specific Embryonic Antigen-4 (SSEA-4), a carbohydrate epitope on the major ganglioside, but not the neutral glycolipid, of human teratocarcinoma cells. As its name implies, the expression of SSEA-4 is stage-specific and can be used to characterize embryonic cells and monitor their differentiation. However, its expression pattern differs in the human and mouse. In the human, SSEA-4 is found on teratocarcinoma (embryonal carcinoma or EC), embryonic inner cell mass (ICM), embryonic stem (ES) cells, and the K562 erythromyeloid leukeumia cell line. As human stem cells undergo differentiation, SSEA-4 expression is lost. In the mouse, SSEA-4 is found on oocytes and early cleavage-stage embryos, and primitive ectoderm, but not on EC, ICM, or ES cells. In some cases, SSEA-4 expression appears upon differentiation of mouse EC or ES cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SSEA-4
Monoclonal
PerCP-Cyanine5.5
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Teratocarcinoma Cell Line
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MC813-70
Stage-Specific Embryonic Antigen-4
Mouse
Affinity Purified
RUO
Human, Mouse
IgG3 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.