missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRR, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SRR protein.
Brand: Abnova H00063826-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MCAQYCISFADVEKAHINIRDSIHLTPVLTSSILNQLTGRNLFFKCELFQKTGSFKIRGALNAVRSLVPDALERKPKAVVTHSSGNHGQALTYAAKLEGIPAYIVVPQTAPDCKKLAIQAYGASIVYCEPSDESRENVAKRVTEETEGIMVHPNQEPAVIAGQGTIALEVLNQVPLVDALVVPVGGGGMLAGIAITVKALKPSVKVYAAEPSNADDCYQSKLKGKLMPNLYPPETIADGVKSSIGLNTWPIIRDLVDDIFTVTEDEIKCATQLVWERMKLLIEPTAGVGVAAVLSQHFQTVSPEVKNICIVLSGGNVDLTSSITWVKQAERPASYQSVSVSpecifications
| SRR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human SRR protein. | |
| SRR | |
| ILV1/ISO1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_021947.1 | |
| SRR | |
| SRR (NP_068766.1, 1 a.a. ∼ 340 a.a) full-length human protein. | |
| RUO | |
| 63826 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction