missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRD5A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542550
This item is not returnable.
View return policy
Description
Peptide sequence: MQVQCQQSPV LAGSATLVAL GALALYVAKP SGYGKHTESL KPAATRLPAR Sequence homology: Human: 100%; Pig: 86%; Rabbit: 91%.
This gene encodes a microsomal protein expressed at high levels in androgen-sensitive tissues such as the prostate. The encoded protein is active at acidic pH and is sensitive to the 4-azasteroid inhibitor finasteride. Deficiencies in this gene can result in male pseudohermaphroditism, specifically pseudovaginal perineoscrotal hypospadias (PPSH).
Specifications
| SRD5A2 | |
| Polyclonal | |
| Unconjugated | |
| SRD5A2 | |
| 3-oxo-5-alpha-steroid 4-dehydrogenase 2; 5 alpha-SR2; 5ART2; MGC138457; S5AR 2; SR type 2; SRD5A2; ST5AR2; steroid 5 alpha reductase type2; steroid 5 alpha-reductase 2; steroid 5-alpha-reductase 2; steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2); type 2; 4-ene-3-oxosteroid 5-alpha oxidoreductase; type II 5-alpha reductase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6716 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P31213 | |
| SRD5A2 | |
| Synthetic peptide directed towards the N-terminal of human SRD5A2 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction