missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTBN2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SPTBN2.
Brand: Abnova H00006712-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: LWRFLWEVGEAEAWVREQQHLLASADTGRDLTGALRLLNKHTALRGEMSGRLGPLKLTLEQGQQLVAEGHPGASQASASpecifications
| SPTBN2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SPTBN2. | |
| SPTBN2 | |
| GTRAP41/SCA5 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006946 | |
| SPTBN2 | |
| SPTBN2 (NP_008877, 643 a.a. ∼ 720 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 6712 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction