missing translation for 'onlineSavingsMsg'
Learn More
Learn More
signal peptide peptidase 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SPPL3.
Brand: Abnova H00121665-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: NSNVMVKVATQPADNPLDVLSRKLHLGPNVGRDVPRLSLPGKLVFPSSTGSSpecifications
| signal peptide peptidase 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SPPL3. | |
| UNQ1887 | |
| DKFZp586C1324/IMP2/MDHV1887/MGC126674/MGC126676/MGC90402/PRO4332/PSL4/SPPL3 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 121665 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_139015 | |
| UNQ1887 | |
| SPPL3 (NP_620584, 212 a.a. ∼ 262 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction