Learn More
serine peptidase inhibitor, Kazal type 1, Mouse, Clone: 4D4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SPINK1.
Brand: Abnova H00006690-M01.100ug
Description
The protein encoded by this gene is a trypsin inhibitor, which is secreted from pancreatic acinar cells into pancreatic juice. It is thought to function in the prevention of trypsin-catalyzed premature activation of zymogens within the pancreas and the pancreatic duct. Mutations in this gene are associated with hereditary pancreatitis and tropical calcific pancreatitis. [provided by RefSeq
Sequence: DSLGREAKCYNELNGCTKIYDPVCGTDGNTYPNECVLCFENRKRQTSILIQKSGPCSpecifications
| serine peptidase inhibitor, Kazal type 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC025790 | |
| SPINK1 | |
| SPINK1 (AAH25790, 24 a.a. ∼ 79 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| 4D4 | |
| Mouse monoclonal antibody raised against a partial recombinant SPINK1. | |
| SPINK1 | |
| PCTT/PSTI/Spink3/TATI | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 6690 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.