missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDG8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523195
This item is not returnable.
View return policy
Description
The amino acid sequence used as immunogen is 84% homologous in Bovine and Porcine.
The lysosphingolipid sphingosine 1-phosphate (S1P) regulates cell proliferation, apoptosis, motility, and neurite retraction. Its actions may be both intracellular as a second messenger and extracellular as a receptor ligand. S1P and the structurally related lysolipid mediator lysophosphatidic acid (LPA) signal cells through a set of G protein-coupled receptors known as EDG receptors. Some EDG receptors (e.g., EDG1) are S1P receptors; others (e.g., EDG2) are LPA receptors.
Specifications
| EDG8 | |
| Polyclonal | |
| Unconjugated | |
| S1PR5 | |
| Edg8; Edg-8; endothelial differentiation G-protein-coupled receptor 8; endothelial differentiation, sphingolipid G-protein-coupled receptor, 8; lpB4; Lysophospholipid receptor B4; nerve growth factor-regulated G-protein-coupled receptor 1; Nrg1; NRG-1; S1P receptor 5; S1P receptor Edg-8; S1P(5) receptor; S1P5; S1PR5; Sphingosine 1-phosphate receptor 5; sphingosine 1-phosphate receptor EDG8; sphingosine 1-phosphate receptor Edg-8; sphingosine-1-phosphate receptor 5; sphingosine-1-phosphate receptor LPB4; SPPR-1; SPPR-2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 53637, 60399, 610828, 94226 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.0 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q91X56, Q9H228, Q9JKM5 | |
| S1PR5 | |
| A synthetic peptide corresponding to amino acids 15-55 of human Edg8. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Frog | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction