missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta-3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA146007
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: Hela whole cell extract.
Spectrin beta-3 Polyclonal antibody specifically detects Spectrin beta-3 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Spectrin beta-3 | |
Polyclonal | |
Unconjugated | |
Sptbn2 | |
beta SpIII sigma 1; beta-III spectrin; beta-spectrin 3; glutamate transporter EAAT4-associated protein 41; GTRAP41; KIAA0302; mKIAA0302; SCA5; SCAR14; Spectrin Beta; spectrin beta 3; spectrin beta chain, brain 2; spectrin beta chain, non-erythrocytic 2; spectrin beta III sigma 2; spectrin beta, non-erythrocytic 2; spectrin, beta, non-erythrocytic 2; spectrin, non-erythroid beta chain 2; spectrin-like protein GTRAP41; spinocerebellar ataxia 5 protein; Spnb3; SPNB-3; SPTBN2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20743, 6712 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O15020 | |
Sptbn2 | |
The epitope recognized by this antibody maps to a C-terminal region between residue 2340 and 2390 of spectrin beta 3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |