missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA9, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SPATA9 protein.
Brand: Abnova H00083890-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MPIKPVGWICGQVLKNFSGRIEGIQKAIMDLVDEFKDEFPTILRLSQSNQKREPAQKTSKIRMAIALAKINRATLIRGLNSISRSSKSVAKLLHPQLACRLLELRDISGRLLREVNAPRQPLYNIQVRKGSLFEIISFPAKTALTSIIYASYAALIYLAVCVNAVLKKVKNIFQEEESIRQNREESENCRKAFSEPVLSEPMFAEGEIKAKPYRSLPEKPDISDYPKLLANKQSNNIQVLHSVFDQSAEMNEQISpecifications
| SPATA9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human SPATA9 protein. | |
| SPATA9 | |
| FLJ35906/NYD-SP16 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 83890 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_031952.2 | |
| SPATA9 | |
| SPATA9 (NP_114158.2, 1 a.a. ∼ 254 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction