Learn More
sperm associated antigen 6, Mouse, Clone: 3E3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SPAG6.
Brand: Abnova H00009576-M01.100ug
Description
The correlation of anti-sperm antibodies with cases of unexplained infertility implicates a role for these antibodies in blocking fertilization. Improved diagnosis and treatment of immunologic infertility, as well as identification of proteins for targeted contraception, are dependent on the identification and characterization of relevant sperm antigens. The protein expressed by this gene is recognized by anti-sperm antibodies from an infertile man. This protein localizes to the tail of permeabilized human sperm and contains eight contiguous armadillo repeats, a motif known to mediate protein-protein interactions. Studies in mice suggest that this protein is involved in sperm flagellar motility and maintenance of the structural integrity of mature sperm. Alternatively spliced variants that encode different protein isoforms have been described but the full-length sequences of only two have been determined. [provided by RefSeq
Sequence: MSQRQVLQVFEQYQKARTQFVQMVAELATRPQNIETLQNAGVMSLLRTLLLDVVPTIQQTAALALGRLANYNDDLAEAVVKCDILPQLVYSLAEQNRFYK*Specifications
| sperm associated antigen 6 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_012443 | |
| SPAG6 | |
| SPAG6 (NP_036575, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3E3 | |
| Mouse monoclonal antibody raised against a partial recombinant SPAG6. | |
| SPAG6 | |
| DKFZp434I153/MGC26276/Repro-SA-1/pf16 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 9576 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.