missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526934
This item is not returnable.
View return policy
Description
SP5 Polyclonal Antibody for Western Blot, Flow
SP5 binds to GC boxes promoters elements. SP5 probable transcriptional activator that has a role in the coordination of changes in transcription required to generate pattern in the developing embryo.
Specifications
| SP5 | |
| Polyclonal | |
| Unconjugated | |
| SP5 | |
| SP5; Sp5 transcription factor; trans-acting transcription factor 5; transcription factor Sp5 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 389058, 64406 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 1.7 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6BEB4, Q9JHX2 | |
| SP5 | |
| KLH conjugated synthetic peptide between 369-398 amino acids from the C-terminal region of human SP5. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction