missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sp1 transcription factor, Mouse, Clone: 1A5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SP1.
Brand: Abnova H00006667-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: GTGELDLTATQLSQGANGWQIISSSSGATPTSKEQSGSSTNGSNGSESSKNRTVSGGQYVVAAAPNLQNQQVLTGLPGVMPNIQYQVIPQFQTVDGQQLQFAATGAQVQQSpecifications
| Sp1 transcription factor | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_138473 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6667 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1A5 | |
| Mouse monoclonal antibody raised against a partial recombinant SP1. | |
| SP1 | |
| SP1 | |
| SP1 (NP_612482, 89 a.a. ∼ 198 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human, Mouse | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction