missing translation for 'onlineSavingsMsg'
Learn More

SOX9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

£185.00 - £410.00

Specifications

Classification Polyclonal
Form Purified
Regulatory Status RUO
Target Species Human, Rat, Canine, Feline, Goat, Primate, Sheep
Isotype IgG
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18785813
View Documents
Novus Biologicals
NBP2-24659SS
0.025 mg
£185.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18755323
View Documents
Novus Biologicals
NBP2-24659
£410.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SOX9 Polyclonal specifically detects SOX9 in Human, Mouse, Rat, Canine, Feline, Goat, Primate, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Polyclonal
RUO
IgG
Purified
Human, Rat, Canine, Feline, Goat, Primate, Sheep
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.