missing translation for 'onlineSavingsMsg'
Learn More

SRY (sex determining region Y)-box 8, Mouse, Clone: 8D8, Abnova™

Mouse monoclonal antibody raised against a partial recombinant SOX8.

Brand:  Abnova H00030812-M02.100ug

Product Code. 16166286

  • £303.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. This protein may be involved in brain development and function. Haploinsufficiency for this protein may contribute to the mental retardation found in haemoglobin H-related mental retardation (ART-16 syndrome). [provided by RefSeq

Sequence: VKRPMNAFMVWAQAARRKLADQYPHLHNAELSKTLGKLWRLLSESEKRPFVEEAERLRVQHKKDHPDYKYQPRRRKSAKAGHSDSDSGAELGPHPGGGAVYKAEAGLGD
Specifications

Specifications

SRY (sex determining region Y)-box 8
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_014587
SOX8
SOX8 (NP_055402, 102 a.a. ∼ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Transcription Regulation
Primary
Human
Antibody
ELISA, Western Blot
8D8
Mouse monoclonal antibody raised against a partial recombinant SOX8.
SOX8
MGC24837
Mouse
Affinity Purified
RUO
Yes
30812
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.