Learn More
SRY (sex determining region Y)-box 6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SOX6.
Brand: Abnova H00055553-A01.50uL
Description
This gene encodes a member of the D subfamily of sex determining region y-related transcription factors that are characterized by a conserved DNA-binding domain termed the high mobility group box and by their ability to bind the minor groove of DNA. The encoded protein is a transcriptional activator that is required for normal development of the central nervous system, chondrogenesis and maintenance of cardiac and skeletal muscle cells. The encoded protein interacts with other family members to cooperatively activate gene expression. Alternative splicing results in multiple transcript variants
Sequence: MSSKQATSPFACAADGEDAMTQDLTSREKEEGSDQHVASHLPLHPIMHNKPHSEELPTLVSTIQQDADWDSVLSSQQRMESENNKLCSLYSFRNTSTSPHKPDEGSRDRSpecifications
| SRY (sex determining region Y)-box 6 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SOX6. | |
| SOX6 | |
| HSSOX6 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_033326 | |
| SOX6 | |
| SOX6 (NP_201583, 1 a.a. ∼ 109 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 55553 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.