Learn More
SRY (sex determining region Y)-box 30, Mouse, Clone: 6H1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SOX30.
Brand: Abnova H00011063-M02.100ug
Description
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may be involved in the differentiation of developing male germ cells. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: SMPECLSYYEDRYPKHEGIFSTLNRDYSFRDYSSECTHSENSRSCENMNGTSYYNSHSHSGEENLNPVPQLDIGTLENVFTAPTSTPSSIQQVNVTDSDEEEEEKVLRDLSpecifications
| SRY (sex determining region Y)-box 30 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_178424 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 11063 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 6H1 | |
| Mouse monoclonal antibody raised against a partial recombinant SOX30. | |
| SOX30 | |
| SOX30 | |
| SOX30 (NP_848511, 644 a.a. ∼ 753 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.