Learn More
SRY (sex determining region Y)-box 21, Mouse, Clone: 3H4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SOX21.
Brand: Abnova H00011166-M02.100ug
Description
SRY-related HMG-box (SOX) genes encode a family of DNA-binding proteins containing a 79-amino acid HMG (high mobility group) domain that shares at least 50% sequence identity with the DNA-binding HMG box of the SRY protein (MIM 480000). SOX proteins are divided into 6 subgroups based on sequence similarity within and outside of the HMG domain. For additional background information on SOX genes, see SOX1 (MIM 602148).[supplied by OMIM
Sequence: KFAFPVPYGLGGVADAEHPALKAGAGLHAGAGGGLVPESLLANPEKAAAAAAAAAARVFFPQSAAAAAAAAAAAAAGSPYSLLDLGSKMAEISSSpecifications
| SRY (sex determining region Y)-box 21 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_007084 | |
| SOX21 | |
| SOX21 (NP_009015, 91 a.a. ∼ 184 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3H4 | |
| Mouse monoclonal antibody raised against a full length recombinant SOX21. | |
| SOX21 | |
| SOX25 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 11166 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.