missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535167
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SOX18 Polyclonal antibody specifically detects SOX18 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
SOX18 | |
Polyclonal | |
Unconjugated | |
SOX18 | |
AI385749; HLTRS; HLTS; Ra; ragged; Ragl; SOX18; Sox-18; SRY (sex determining region Y)-box 18; SRY box 18; SRY-box 18; SRY-box containing gene 18; SRY-box transcription factor 18; Sry-related HMG-box gene 18; transcription factor SOX-18 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
54345 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P35713 | |
SOX18 | |
KLH conjugated synthetic peptide between 59-88 amino acids from the N-terminal region of human SOX18 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |