missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX17 Rabbit anti-Human, Marsupial, Mouse, Non-human primate, Rat, Zebrafish, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523352
This item is not returnable.
View return policy
Description
SOX17, also known as transcription factor SOX-17, as the name suggests is a transcription factor or a probable transcriptional activator in the premeiotic germ cells. It belongs to the SRY-related HMG box family of transcription factors with a high-mobility group DNA-binding domain (HMG box), that plays a key role in embryogenesis and binds to the sequences 5′-AACAAT-′3 or 5′-AACAAAG-3′. SOX17 plays an essential role in cardiac mesoderm specification as well as in definitive hematopoiesis, maintaining fetal hematopoietic stem cells, hematopoiesis in the fetal liver and yolk sac. Along with beta-catenin, it is known to cooperate in the regulation of transcription of endodermal genes. Defects in SOX17 are known to cause a severe defects in fetal hematopoiesis.
Specifications
| SOX17 | |
| Polyclonal | |
| Unconjugated | |
| Q9H6I2, Q61473 | |
| SOX17 | |
| Synthetic peptide (80 KDERKRLAQQNPDLHNAEL 98) of SOX17 protein. | |
| 100 μg | |
| Primary | |
| Human, Marsupial, Mouse, Non-human Primate, Rat, Zebrafish | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| SOX17 | |
| VUR3, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20671, 30544, 312936, 64321 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction