Learn More
Sox17 Mouse anti-Human, PerCP-Cy5.5, Clone: P7-969, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562387
Description
The P7-969 monoclonal antibody reacts with human Sox17, a member of the SOX (SRY-releated HMG-box) family of transcription factors. SOX family members contain a DNA binding domain (HMG-box) and are involved in the control of development. Sox17 is expressed in primitive and definitive endoderm and regulates fetal and neonatal hematopoietic stem cell proliferation.
Specifications
| Sox17 | |
| Monoclonal | |
| PerCP-Cyanine5.5 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Sox17 recombinant protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| P7-969 | |
| SOX-17, SOX17, FLJ22252 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.