missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX11 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA537891
Code : NEW
Additional Details : Weight : 0.00900kg
Description
This antibody is predicted to react with mouse, rat, pig and cow based on sequence homology.
SOX11 Polyclonal antibody specifically detects SOX11 in Human samples. It is validated for Western Blot.Specifications
SOX11 | |
Polyclonal | |
Unconjugated | |
SOX11 | |
1110038H03Rik; 6230403H02Rik; AI836553; end1; MRD27; Sox11; Sox-11; SRY; SRY (sex determining region Y)-box 11; SRY (sex-determining region Y)-box 11; SRY-box 11; SRY-box containing gene 11; SRY-box transcription factor 11; SRY-related HMG-box gene 11; transcription factor SOX-11 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6664 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P35716 | |
SOX11 | |
Peptide with sequence C-SRLYYSFKNITKQHP, from the internal region of SOX11 (aa309-323) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |