Learn More
SNIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA517309
Product Code. 11526131
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
It is not recommended to aliquot this antibody.
Acts as a negative regulator of SRC by activating CSK which inhibits SRC activity and downstream signaling, leading to impaired cell spreading and migration. Regulates dendritic spine morphology. Involved in calcium-dependent exocytosis. May play a role in neurotransmitter release or synapse maintenance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SNIP
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Q9C0H9, Q9QWI6, Q9QXY2
SRCIN1
Synthetic peptide corresponding to residues near the carboxy terminus of human SNIP/p140Cap
100 μL
Primary
Human, Mouse, Non-human Primate, Rat
Antibody
IgG
Western Blot
Unconjugated
SRCIN1
KIAA1684; mKIAA1684; p130Cas-associated protein; P140; p140Cap; SNAP-25-interacting protein; SNAP25-interacting protein; SNIP; SRC kinase signaling inhibitor 1; SRCIN1
Rabbit
Antigen affinity chromatography
RUO
56013, 56029, 80725
-20°C