missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAIL Monoclonal Antibody (F.31.8)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514801
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Zinc finger protein SNAI1 (SNAIL)is a zinc finger transcriptional repressor which down regulates the expression of the ectodermal gene within the mesoderm and functions to form the mesoderm and neural crest. It is reported that SNAI1 acts as an important factor of tumor invasion as evidenced by its role in E-cadherin down-regulation and induction of epithelial-mesenchymal transition (EMT). In human breast cancer, the expression of SNAI1 and/or the homologous SNAI2 (Slug) has been associated with E-cadherin repression, local or distant metastasis, tumor recurrence, or poor prognosis in different tumor series. It is also reported that Snail 1 protein in the stromamay be anew putative prognosis marker for colon tumors.
Specifications
| SNAIL | |
| Monoclonal | |
| 110 μg/mL | |
| 0.01M HEPES with 50% glycerol, 0.15M NaCl, 100 μg/mL BSA and <0.02% sodium azide, pH 7.5 | |
| O95863, Q02085 | |
| Snai1 | |
| Recombinant human Snail protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| F.31.8 | |
| Unconjugated | |
| Snai1 | |
| AI194338; dJ710H13.1; Protein sna; protein snail homolog 1; SLUGH2; SNA; Sna protein; Sna1; SNAH; snai; SNAI1; Snail; snail 1; snail 1 homolog; snail 1 zinc finger protein; snail 1, zinc finger protein; snail family transcriptional repressor 1; snail family zinc finger 1; snail homolog 1; Snail1; Zinc finger protein SNAI1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116490, 20613, 6615 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction