missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAI3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531970
This item is not returnable.
View return policy
Description
Recommended positive controls: A549. Predicted reactivity: Rhesus Monkey (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SNAI3 is a member of the SNAIL gene family, named for the Drosophila snail gene, which plays roles in mesodermal formation during embryogenesis (Katoh and Katoh, 2003).
Specifications
| SNAI3 | |
| Polyclonal | |
| Unconjugated | |
| SNAI3 | |
| AI643946; Protein snail homolog 3; Smuc; SNAI3; snail family transcriptional repressor 3; snail family zinc finger 3; snail homolog 3; SNAIL3; snail-related gene from muscle cells; snail-related protein 2; Zfp293; zinc finger protein 293; Zinc finger protein SNAI3; ZNF293 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 333929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q3KNW1 | |
| SNAI3 | |
| Recombinant protein encompassing a sequence within the center region of human SNAI3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction