missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAI1, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00006615-PW1.1Set
Includes: Antibody pair for IP: mouse monoclonal anti-SNAI1 (300μg), Antibody pair for WB: rabbit polyclonal anti-SNAI1 (50μL)
Specifications
| SNAI1 | |
| Monoclonal/Polyclonal | |
| This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
| SLUGH2/SNA/SNAH/dJ710H13.1 | |
| Mouse, Rabbit | |
| RUO | |
| 6615 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| SNAI1 | |
| SNAI1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-SNAI1 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-SNAI1 (50 ul) | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction