missing translation for 'onlineSavingsMsg'
Learn More

SUMO-3 Rabbit anti-Equine, Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Product Code. 13217718
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13217718 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13217718 Supplier Invitrogen Supplier No. PA523110

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SUMO3 (SMT3) is a Ubiquitin-like protein found in Saccharomyces cerevisiae similar to the mammalian Ubiquitin-like SUMO1, which is involved in posttranslational protein modification. SUMO3 is functionally active in regulation of nucleocytoplasmic transport and suggest a role in cell-cycle regulation. Knockdown of SUMO3 in Drosophila in the prothoracic gland, which controls key developmental processes through the synthesis and release of ecdysteroids, caused a 4-fold prolongation of larval life and completely blocked the transition from larval to pupal stages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO-3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Gene SUMO3
Gene Accession No. P55854, Q9Z172
Gene Alias SMT3A, SMT3H1, SUMO-3, Smt3B, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SUMO-2, small ubiquitin-related modifier 3, ubiquitin-like protein SMT3A, ubiquitin-like protein SMT3B
Gene Symbols SUMO3
Host Species Rabbit
Immunogen Synthetic peptide (71 CDTPAQLEMEDE 81) of SMT3/SUMO 3 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100630015, 20610, 6612
Target Species Equine, Human, Mouse, Non-human Primate
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.