missing translation for 'onlineSavingsMsg'
Learn More
Learn More
smoothened homolog (Drosophila), Mouse, Clone: 1D9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SMO.
Brand: Abnova H00006608-M11.100ug
This item is not returnable.
View return policy
Description
Sequence: NLWLVEAEISPELQKRLGRKKKRRKRKKEVCPLAPPPELHPPAPAPSTIPRLPQLPRQKCLVAAGAWGAGDSCRQGAWTLVSNPFCPEPSPPQDPFLPSAPAPVAWAHGRRQGLGPIHSRTNLMDTELMDADSDFSpecifications
| smoothened homolog (Drosophila) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005631 | |
| SMO | |
| SMO (NP_005622.1, 653 a.a. ∼ 787 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1D9 | |
| Mouse monoclonal antibody raised against a full-length recombinant SMO. | |
| SMO | |
| Gx/SMOH | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6608 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction