missing translation for 'onlineSavingsMsg'
Learn More
Learn More
structural maintenance of chromosomes 4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SMC4L1.
Brand: Abnova H00010051-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: DSIDIAQECVNFLKRQNIGVATFIGLDKMAVWAKKMTEIQTPENTPRLFDLVKVKDEKIRQAFYFALRDTLVADNLDQATRVAYQKDRRWRVVTLQGQIIEQSGTMTGGGSpecifications
| structural maintenance of chromosomes 4 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SMC4L1. | |
| SMC4 | |
| CAPC/SMC4L1/hCAP-C | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005496 | |
| SMC4 | |
| SMC4L1 (NP_005487, 646 a.a. ∼ 755 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 10051 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction