Learn More
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2, Mouse, Clone: 2B2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SMARCD2.
Brand: Abnova H00006603-M02.100ug
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: RDFMLSFSTDPQDFIQEWLRSQRRDLKIITDVIGNPEEERRAAFYHQPWAQEAVGRHIFAKVQQRRQELEQVLGIRLSpecifications
| SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003077 | |
| SMARCD2 | |
| SMARCD2 (NP_003068, 398 a.a. ∼ 474 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2B2 | |
| Mouse monoclonal antibody raised against a partial recombinant SMARCD2. | |
| SMARCD2 | |
| BAF60B/CRACD2/PRO2451/Rsc6p | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6603 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.