Learn More
Smad1/Smad8 Rat, PE, Clone: N6-1233, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562509
Description
The N6-1233 monoclonal antibody specifically binds to the Smad1 protein phosphorylated at the Ser463/465 sites and the Smad8 protein phosphorylated at the Ser465/467 sites. Smad1 and Smad8 are ∽60 kDa proteins and are members of the Smad superfamily. The Smad family members are divided into three subfamilies: receptor regulated Smads or R-Smads, including Smads1, 2, 3, 5, and 8; common partner Smad, or Co-Smad, including Smad4; and inhibitory Smads, or I-Smad, including Smads 6 and 7. Activation of Transforming Growth Factor-beta (TGF-beta) superfamily serine/threonine kinase receptors, such as TGF-beta and Bone MorphogenicProtein (BMP) receptors, leads to the phosphorylation of R-Smads at several sites. It has been shown that Ser463 and Ser465 of Smad1 are phosphorylated by BMP receptors. In B cells and pre-B cells, BMP-6 has been shown to induce Smad1/5/8 phosphorylation and inhibit cell proliferation. Phosphorylated R-Smads form complexes with Co-Smad and translocate into the nucleus to regulate transcription affecting a wide range of critical processes including cell-fate determination, proliferation, morphogenesis, differentiation and apoptosis. The inhibitory Smads inhibit this pathway through two potential mechanisms: either by preventing R-Smads from binding to their corresponding receptors and/or by competing with Smad4, the Co-Smad, from binding to R-Smads. This antibody may crossreact with Smad5 pS463/pS465 based on sequence homology.
Specifications
| Smad1/Smad8 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated human Smad1 aa 456-465 peptide | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| N6-1233 | |
| Smad1,Smad8/Smad9; MADH1, MADH9 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.