Learn More
secreted LY6/PLAUR domain containing 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SLURP1 protein.
Brand: Abnova H00057152-D01P.100ug
Description
The protein encoded by this gene is a member of the Ly6/uPAR family but lacks a GPI-anchoring signal sequence. It is thought that this secreted protein contains antitumor activity. Mutations in this gene have been associated with Mal de Meleda, a rare autosomal recessive skin disorder. This gene maps to the same chromosomal region as several members of the Ly6/uPAR family of glycoprotein receptors. [provided by RefSeq
Sequence: MASRWAVQLLLVAAWSMGCGEALKCYTCKEPMTSASCRTITRCKPEDTACMTTLVTVEAEYPFNQSPVVTRSCSSSCVATDPDSIGAAHLIFCCFRDLCNSELSpecifications
| secreted LY6/PLAUR domain containing 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human SLURP1 protein. | |
| SLURP1 | |
| ANUP/ARS/ArsB/LY6LS/MDM | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_020427 | |
| SLURP1 | |
| SLURP1 (NP_065160.1, 1 a.a. ∼ 103 a.a) full-length human protein. | |
| 100 μg | |
| Cytokines | |
| 57152 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.