missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLIT2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLIT2.
Brand: Abnova H00009353-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: GWMGPLCDQRTNDPCLGNKCVHGTCLPINAFSYSCKCLEGHGGVLCDEEEDLFNPCQAIKCKHGKCRLSGLGQPYCECSSGYTGDSCDREISCRGERIRDSpecifications
| SLIT2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SLIT2. | |
| SLIT2 | |
| FLJ14420/SLIL3/Slit-2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004787 | |
| SLIT2 | |
| SLIT2 (NP_004778, 1361 a.a. ∼ 1460 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 9353 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction