Learn More
SLIT2 Rabbit anti-Human, Mouse, Clone: SP122, Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA516423
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is prostate adenocarcinoma.
SLIT2 are ligands for Robo receptors and play a vital role for axonal migration/navigation during neural development. A number of cleavage products through alternate splicing are reported in the literature for SLIT2 proteins. The C-terminal cleavage proteins are more diffusible than the larger N-terminal protein that is more tightly cell associated. SLIT2 protein is expressed in tissues such as fetal lung, kidney, and adult spinal cord.
Specifications
| SLIT2 | |
| Monoclonal | |
| Unconjugated | |
| Q9R1B9, O94813 | |
| SLIT2 | |
| Synthetic peptide corresponding to C-terminus of human SLIT2 protein. | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| SP122 | |
| SLIT2 | |
| SLIL3, Slit-2, slit homolog 2 protein | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 20563, 9353 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.