missing translation for 'onlineSavingsMsg'
Learn More
Learn More
solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC35A3.
Brand: Abnova H00023443-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: DSKCSLRALNRVLHDEILNKPMETLKLAIPSGIYTLQNNLLYVALSNLDAATYSpecifications
| solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SLC35A3. | |
| SLC35A3 | |
| DKFZp781P1297 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 23443 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_012243 | |
| SLC35A3 | |
| SLC35A3 (NP_036375, 61 a.a. ∼ 113 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction