Learn More
solute carrier family 26, member 9, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC26A9.
Brand: Abnova H00115019-A01.50uL
Description
This gene is one member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures yet have markedly different tissue expression patterns. The product of this gene is a highly selective chloride ion channel regulated by WNK kinases. Alternative splicing results in multiple transcript variants encoding differing isoforms
Sequence: QTQFRNGYALAQVMDTDIYVNPKTYNRAQDIQGIKIITYCSPLYFANSEIFRQKVIAKTGMDPQKVLLAKQKYLKKQEKRRMRPTQQRRSLFMKTKTVSLQELQQDFENASpecifications
| solute carrier family 26, member 9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SLC26A9. | |
| SLC26A9 | |
| SLC26A9 | |
| SLC26A9 (NP_443166, 496 a.a. ∼ 605 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_052934 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 115019 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.