Learn More
solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SLC11A1 protein.
Brand: Abnova H00006556-B01P.50ug
Description
This gene is a member of the solute carrier family 11 (proton-coupled divalent metal ion transporters) family and encodes a multi-pass membrane protein. The protein functions as a divalent transition metal (iron and manganese) transporter involved in iron metabolism and host resistance to certain pathogens. Mutations in this gene have been associated with susceptibility to infectious diseases such as tuberculosis and leprosy, and inflammatory diseases such as rheumatoid arthritis and Crohn disease. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined. [provided by RefSeq
Sequence: MTGDKGPQRLSGSSYGSISSPTSPTSPGPQQAPPRETYLRNIESDLQAGAVAGFKLLWVLLWATVLGLLCQRLAARLGVVTGKDLGEVCHLYYPKVPRTVLWLTIELAIVGSDMQEVIGTAIAFNLLSAGRYHPSVPQLFRPGREQLLLLPPLTSPSQSLFYPAVPSEAGLLPCFPEMSpecifications
| solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human SLC11A1 protein. | |
| SLC11A1 | |
| LSH/NRAMP/NRAMP1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 6556 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC041787.1 | |
| SLC11A1 | |
| SLC11A1 (AAH41787.1, 1 a.a. ∼ 178 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.