missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAMF8 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA537956
Code : NEW
Additional Details : Weight : 0.00900kg
Description
SLAMF8 Polyclonal antibody specifically detects SLAMF8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SLAMF8 | |
Polyclonal | |
Unconjugated | |
Slamf8 | |
5830408F06Rik; B lymphocyte activator macrophage expressed; BCM-like membrane protein; Blame; B-lymphocyte activator macrophage expressed; CD353; FLJ20442; MGC129578; SBBI42; SLAM family member 8; SLAMF8 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
56833 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9P0V8 | |
Slamf8 | |
Peptide with sequence C-HADRVGPETENP, from the C-terminus of SLAMF8 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |