missing translation for 'onlineSavingsMsg'
Learn More

SIII p15 Mouse, Unlabeled, Clone: 56, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610760

 View more versions of this product

Product Code. 15845629

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

SIII p15 is one of three polypeptides that comprise the approximate 140kDa RNA polymerase II transcription factor SIII. The other two polypeptides have the molecular weights of 110kDa and 18kDa. All three polypeptides are required for transcription factor SIII function. Although the polypeptides were identified and purified from rat liver nuclear extracts, tissue expression of this transcription factor appears to be widespread. RNA polymerase II recognizes the SIII transcription factor and in turn initiates transcription from these correctly initiated transcripts. A yeast transcription factor designated YES may be the yeast homolog to SIII.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIII p15
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human SIII p15 aa. 1-112
50 μg
Cell Biology
Canine, Frog, Human, Mouse, Rat
IgG1
Western Blot
56
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.